Merck
  • Towards the differential diagnosis of prostate cancer by the pre-treatment of human urine using ionic liquids.
Towards the differential diagnosis of prostate cancer by the pre-treatment of human urine using ionic liquids.
Scientific reports (2020-09-12)
Matheus M Pereira, João D Calixto, Ana C A Sousa, Bruno J Pereira, Álvaro S Lima, João A P Coutinho, Mara G Freire
ZUSAMMENFASSUNG
Prostate specific antigen (PSA) is the most widely used clinical biomarker for the diagnosis and monitoring of prostate cancer. Most available techniques for PSA quantification in human fluids require extensive sample processing and expensive immunoassays that are often unavailable in developing countries. The quantification of PSA in serum is the most common practice; however, PSA is also present in human urine, although less used in diagnosis. Herein we demonstrate the use of ionic-liquid-based aqueous biphasic systems (IL-based ABS) as effective pre-treatment strategies of human urine, allowing the PSA detection and quantification by more expedite equipment in a non-invasive matrix. If properly designed, IL-based ABS afford the simultaneous extraction and concentration of PSA (at least up to 250-fold) in the IL-rich phase. The best ABS not only allow to concentrate PSA but also other forms of PSA, which can be additionally quantified, paving the way to their use in differential prostate cancer diagnosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrabutylphosphoniumhydroxid -Lösung, 40 wt. % in H2O
Sigma-Aldrich
1,3-Propylene sulfite, 99%