Merck
  • Engineering of Synechococcus sp. strain PCC 7002 for the photoautotrophic production of light-sensitive riboflavin (vitamin B2).
Engineering of Synechococcus sp. strain PCC 7002 for the photoautotrophic production of light-sensitive riboflavin (vitamin B2).
Metabolic engineering (2020-09-30)
Benjamin Kachel, Matthias Mack
ZUSAMMENFASSUNG
Due to their capability of photosynthesis and autotrophic growth, cyanobacteria are currently investigated with regard to the sustainable production of a wide variety of chemicals. So far, however, no attempt has been undertaken to engineer cyanobacteria for the biotechnological production of vitamins, which is probably due to the light-sensitivity of many of these compounds. We now describe a photoautotrophic bioprocess to synthesize riboflavin, a vitamin used as a supplement in the feed and food industry. By overexpressing the riboflavin biosynthesis genes ribDGEABHT from Bacillus subtilis in the marine cyanobacterium Synechococcus sp. PCC 7002 riboflavin levels in the supernatant of the corresponding recombinant strain increased 56-fold compared to the wild-type. Introduction of a second promoter region upstream of the heterologous ribAB gene - coding for rate-limiting enzymatic functions in the riboflavin biosynthesis pathway - led to a further increase of riboflavin levels (211-fold compared to the wild-type). Degradation of the light-sensitive product riboflavin was prevented by culturing the genetically engineered Synechococcus sp. PCC 7002 strains in the presence of dichromatic light generated by red light-emitting diodes (λ = 630 and 700 nm). Synechococcus sp. PCC 7002 naturally is resistant to the toxic riboflavin analog roseoflavin. Expression of the flavin transporter pnuX from Corynebacterium glutamicum in Synechococcus sp. PCC 7002 resulted in roseoflavin-sensitive recombinant strains which in turn could be employed to select roseoflavin-resistant, riboflavin-overproducing strains as a chassis for further improvement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridiniumchlorchromat, 98%
Sigma-Aldrich
Lumichrom