Merck
  • New insights into the fractionation of effluent organic matter on diagnosis of key composition affecting advanced phosphate removal by Zr-based nanocomposite.
New insights into the fractionation of effluent organic matter on diagnosis of key composition affecting advanced phosphate removal by Zr-based nanocomposite.
Water research (2020-08-28)
Bin Lin, Yanyang Zhang, Feifan Shen, Lu Zhang, Dan Wang, Xiaobo Tang, Yong Zhou, Xinyu Nie, Lu Lv, Weiming Zhang, Ming Hua, Bingcai Pan
ZUSAMMENFASSUNG
The influence of effluent organic matter (EfOM) on phosphate removal by adsorption plays an important role in evaluating the applicability of adsorbents. Currently, molecular understanding of EfOM regarding its impact on adsorption is insufficient due to a lack of appropriate EfOM fractionation/characterization protocols, as associated with the specific structure-function property of adsorbents. In this work, a combined method coupling DEAE/XAD fractionation with molecular characterization was proposed, targeting the versatile structure-function characters of nanocomposite, to reveal the composition of EfOM as well as its impact on phosphate removal by nanocomposite during long-term adsorption/regeneration runs. Zirconium-based polystyrene anion exchanger (HZO-201) was selected as a representative nanocomposite, featuring with porous networking matrix, positively charged surface and multiple adsorptive sites. The EfOM samples from three biologically treated sewage effluent sources were separated into fractions of negatively charged organic acid (OA) and hydrophobic-, transphilic-, hydrophilic-neutral/base (HPO-n/b, TPI-n/b and HPI-n/b). The combined method enables effective differentiation of the charge, aromaticity, molecular weight and functionalities of the fractions, matching the multiple structural/surface characteristics of HZO-201 and favoring the evaluation on the impact of the EfOM fractions. The interference sequence of the EfOM fractions on phosphate removal followed an order of OA > HPO-n/b > TPI-n/b > HPI-n/b. The OA fraction, characterized by negatively charged, aromatic functionalities and broad molecular weight distribution (1-5 kDa and 14 kDa), was recognized as the key interfering fraction, presumably due to its multiple adsorption pathways (i.e., ion exchange, π-π interactions and pore filling). Particularly, the low-molecular-weight OA moieties (1-4 kDa) progressively accumulated onto the nanocomposite via irreversible adsorption, causing a continuous phosphate-capacity loss by 32.70% over multiple cycles. We believe the combined fractionation/characterization method may be extended to other complex water systems to identify key influential organic matters in polishing treatment of various pollutants by adsorption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Amberlite XAD4, 20-60 mesh