Merck
Dissecting transcriptomic signatures of neuronal differentiation and maturation using iPSCs.
Nature communications (2020-01-25)
Emily E Burke, Joshua G Chenoweth, Joo Heon Shin, Leonardo Collado-Torres, Suel-Kee Kim, Nicola Micali, Yanhong Wang, Carlo Colantuoni, Richard E Straub, Daniel J Hoeppner, Huei-Ying Chen, Alana Sellers, Kamel Shibbani, Gregory R Hamersky, Marcelo Diaz Bustamante, BaDoi N Phan, William S Ulrich, Cristian Valencia, Amritha Jaishankar, Amanda J Price, Anandita Rajpurohit, Stephen A Semick, Roland W Bürli, James C Barrow, Daniel J Hiler, Stephanie C Page, Keri Martinowich, Thomas M Hyde, Joel E Kleinman, Karen F Berman, Jose A Apud, Alan J Cross, Nicholas J Brandon, Daniel R Weinberger, Brady J Maher, Ronald D G McKay, Andrew E Jaffe
ZUSAMMENFASSUNG
Human induced pluripotent stem cells (hiPSCs) are a powerful model of neural differentiation and maturation. We present a hiPSC transcriptomics resource on corticogenesis from 5 iPSC donor and 13 subclonal lines across 9 time points over 5 broad conditions: self-renewal, early neuronal differentiation, neural precursor cells (NPCs), assembled rosettes, and differentiated neuronal cells. We identify widespread changes in the expression of both individual features and global patterns of transcription. We next demonstrate that co-culturing human NPCs with rodent astrocytes results in mutually synergistic maturation, and that cell type-specific expression data can be extracted using only sequencing read alignments without cell sorting. We lastly adapt a previously generated RNA deconvolution approach to single-cell expression data to estimate the relative neuronal maturity of iPSC-derived neuronal cultures and human brain tissue. Using many public datasets, we demonstrate neuronal cultures are maturationally heterogeneous but contain subsets of neurons more mature than previously observed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Y-27632 -dihydrochlorid, ≥98% (HPLC)