Merck
  • Regulatory T Cells Expressing Tumor Necrosis Factor Receptor Type 2 Play a Major Role in CD4+ T-Cell Impairment During Sepsis.
Regulatory T Cells Expressing Tumor Necrosis Factor Receptor Type 2 Play a Major Role in CD4+ T-Cell Impairment During Sepsis.
The Journal of infectious diseases (2020-07-23)
Benjamin J Gaborit, Antoine Roquilly, Cédric Louvet, Abderrahmane Sadek, Benoit Tessoulin, Alexis Broquet, Cédric Jacqueline, Mickael Vourc'h, Tanguy Chaumette, Marie Chauveau, Antoine Asquier, Alexandre Bourdiol, Virginie Le Mabecque, Marion Davieau, Jocelyne Caillon, David Boutoille, Fanny Coulpier, Sophie Lemoine, Emilie Ronin, Jérémie Poschmann, Benoit L Salomon, Karim Asehnoune
ZUSAMMENFASSUNG
Sepsis causes inflammation-induced immunosuppression with lymphopenia and alterations of CD4+ T-cell functions that renders the host prone to secondary infections. Whether and how regulatory T cells (Treg) are involved in this postseptic immunosuppression is unknown. We observed in vivo that early activation of Treg during Staphylococcus aureus sepsis induces CD4+ T-cell impairment and increases susceptibility to secondary pneumonia. The tumor necrosis factor receptor 2 positive (TNFR2pos) Treg subset endorsed the majority of effector immunosuppressive functions, and TNRF2 was particularly associated with activation of genes involved in cell cycle and replication in Treg, probably explaining their maintenance. Blocking or deleting TNFR2 during sepsis decreased the susceptibility to secondary infection. In humans, our data paralleled those in mice; the expression of CTLA-4 was dramatically increased in TNFR2pos Treg after culture in vitro with S. aureus. Our findings describe in vivo mechanisms underlying sepsis-induced immunosuppression and identify TNFR2pos Treg as targets for therapeutic intervention.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)