Merck
Distinct differences in chromatin structure at subtelomeric X and Y' elements in budding yeast.
PloS one (2009-07-25)
Xuefeng Zhu, Claes M Gustafsson
ZUSAMMENFASSUNG
In Saccharomyces cerevisiae, all ends of telomeric DNA contain telomeric repeats of (TG(1-3)), but the number and position of subtelomeric X and Y' repeat elements vary. Using chromatin immunoprecipitation and genome-wide analyses, we here demonstrate that the subtelomeric X and Y' elements have distinct structural and functional properties. Y' elements are transcriptionally active and highly enriched in nucleosomes, whereas X elements are repressed and devoid of nucleosomes. In contrast to X elements, the Y' elements also lack the classical hallmarks of heterochromatin, such as high Sir3 and Rap1 occupancy as well as low levels of histone H4 lysine 16 acetylation. Our analyses suggest that the presence of X and Y' elements govern chromatin structure and transcription activity at individual chromosome ends.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nuclease mikrokokkal aus Staphylococcus aureus, 100-300 units/mg protein
Sigma-Aldrich
Nuclease mikrokokkal aus Staphylococcus aureus, 100-300 units/mg protein
Sigma-Aldrich
Anti-Histon H4-Antikörper, pan, Klon 62-141-13, Kaninchen, monoklonal, clone 62-141-13, Upstate®, from rabbit