Merck
Expansion of functional personalized cells with specific transgene combinations.
Nature communications (2018-03-10)
Christoph Lipps, Franziska Klein, Tom Wahlicht, Virginia Seiffert, Milada Butueva, Jeannette Zauers, Theresa Truschel, Martin Luckner, Mario Köster, Roderick MacLeod, Jörn Pezoldt, Jochen Hühn, Qinggong Yuan, Peter Paul Müller, Henning Kempf, Robert Zweigerdt, Oliver Dittrich-Breiholz, Thomas Pufe, Rainer Beckmann, Wolf Drescher, Jose Riancho, Carolina Sañudo, Thomas Korff, Bertram Opalka, Vera Rebmann, Joachim R Göthert, Paula M Alves, Michael Ott, Roland Schucht, Hansjörg Hauser, Dagmar Wirth, Tobias May
ZUSAMMENFASSUNG
Fundamental research and drug development for personalized medicine necessitates cell cultures from defined genetic backgrounds. However, providing sufficient numbers of authentic cells from individuals poses a challenge. Here, we present a new strategy for rapid cell expansion that overcomes current limitations. Using a small gene library, we expanded primary cells from different tissues, donors, and species. Cell-type-specific regimens that allow the reproducible creation of cell lines were identified. In depth characterization of a series of endothelial and hepatocytic cell lines confirmed phenotypic stability and functionality. Applying this technology enables rapid, efficient, and reliable production of unlimited numbers of personalized cells. As such, these cell systems support mechanistic studies, epidemiological research, and tailored drug development.