Merck
  • Interleukin-6 increases adrenal androgen release by regulating the expression of steroidogenic proteins in NCI-H295R cells.
Interleukin-6 increases adrenal androgen release by regulating the expression of steroidogenic proteins in NCI-H295R cells.
Journal of cellular physiology (2020-04-30)
Xian Wei, Junfeng Zhang, Wei Peng, Hao Xu, Zhewen Wei, Linhao Pang, Jihong Liu, Tao Wang
ZUSAMMENFASSUNG
The purpose of this study was to investigate the potential mechanism of interleukin-6 (IL-6) on the stimulation of excessive androgen secretion in human NCI-H295R adrenocortical cells. We performed transcriptome sequencing of cancer and paracancerous tissues obtained from functional adrenal cortical adenomas. The secretion of dehydroepiandrosterone sulfate (DHEAS) in NCI-H295R cells was detected by a chemiluminescence assay. The expression of messenger RNA (mRNA) was detected by real-time polymerase chain reaction and that of protein was detected by western blotting. The expression of secretogranin II (SCG2) and IL-6 were significantly increased in cancer tissues. Upregulation of mRNA and protein levels of AKR1C3, CYP11A, CYP17A1, 3βHSD, and SULT2A1 was observed after stimulation with IL-6. IL-6 could also increase the expression of StAR mRNA and proteins. Our results suggest that IL-6 can promote androgen secretion by regulating the expression of genes related to androgen pathways.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O