Merck
APP anterograde transport requires Rab3A GTPase activity for assembly of the transport vesicle.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-11-20)
Anita Szodorai, Yung-Hui Kuan, Silke Hunzelmann, Ulrike Engel, Ayuko Sakane, Takuya Sasaki, Yoshimi Takai, Joachim Kirsch, Ulrike Müller, Konrad Beyreuther, Scott Brady, Gerardo Morfini, Stefan Kins
ZUSAMMENFASSUNG
The amyloid precursor protein (APP) is anterogradely transported by conventional kinesin in a distinct transport vesicle, but both the biochemical composition of such a vesicle and the specific kinesin-1 motor responsible for transport are poorly defined. APP may be sequentially cleaved by beta- and gamma-secretases leading to accumulation of beta-amyloid (Abeta) peptides in brains of Alzheimer's disease patients, whereas cleavage of APP by alpha-secretases prevents Abeta generation. Here, we demonstrate by time-lapse analysis and immunoisolations that APP is a cargo of a vesicle containing the kinesin heavy chain isoform kinesin-1C, the small GTPase Rab3A, and a specific subset of presynaptic protein components. Moreover, we report that assembly of kinesin-1C and APP in this vesicle type requires Rab3A GTPase activity. Finally, we show cleavage of APP in transport vesicles by alpha-secretase activity, likely mediated by ADAM10. Together, these data indicate that maturation of APP transport vesicles, including recruitment of conventional kinesin, requires Rab3 GTPase activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.1 % (w/v) in H2O
Sigma-Aldrich
Ziegenserum
Sigma-Aldrich
Poly-L-Lysin -Lösung, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Paraformaldehyd, powder, 95%
Sigma-Aldrich
Paraformaldehyd, prilled, 95%
Sigma-Aldrich
Paraformaldehyd, reagent grade, crystalline
Sigma-Aldrich
Paraformaldehyd, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylendiamin