Merck
  • Podocytes present antigen to activate specific T cell immune responses in inflammatory renal disease.
Podocytes present antigen to activate specific T cell immune responses in inflammatory renal disease.
The Journal of pathology (2020-07-21)
Shan Li, Ying Liu, Yueqin He, Weiwei Rong, Mingchao Zhang, Limin Li, Zhihong Liu, Ke Zen
ZUSAMMENFASSUNG
Infiltration of activated T cells into renal tissue plays an essential role in inflammatory nephropathy. However, the mechanism enabling the renal recruitment and activation of T cells remains elusive. Here we report that inflammatory cytokine-promoted antigen presentation by podocytes is a key for recruiting and activating specific T cells. Our results showed that diabetes-associated inflammatory cytokines IFNγ and IL-17 all upregulated expression of MHC-I, MHC-II, CD80 and CD86 on the podocyte surface. Both IFNγ and IL-17 stimulated the uptake and processing of ovalbumin (OVA) by mouse podocytes, resulting in presentation of OVA antigen peptide on the cell surface. OVA antigen presentation by podocytes was also validated using human podocytes. Furthermore, OVA antigen-presenting mouse podocytes were able to activate OT-I mouse T cell proliferation and inflammatory cytokine secretion, which in turn caused podocyte injury and apoptosis. Finally, OT-I mice subjected to direct renal injection of OVA plus IFNγ/IL-17 but not OVA alone exhibited OVA antigen presentation by podocytes and developed nephropathy in 4 weeks. In conclusion, antigen presentation by podocytes under inflammatory conditions plays an important role in activating T cell immune responses and facilitating immune-mediated glomerular disease development. © 2020 Pathological Society of Great Britain and Ireland. Published by John Wiley & Sons, Ltd.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Albumin aus Hühnereiweiss, lyophilized powder, ≥98% (agarose gel electrophoresis)
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Anti-Podocin in Kaninchen hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Protease aus Streptomyces griseus, BioReagent, DNase, RNase, and nickase, none detected (No RNase.)