Merck
  • Antioxidant and antimicrobial activities of different enzymatic hydrolysates from desalted duck egg white.
Antioxidant and antimicrobial activities of different enzymatic hydrolysates from desalted duck egg white.
Asian-Australasian journal of animal sciences (2020-02-15)
Rommanee Thamamsena, Deng Cheng Liu
ZUSAMMENFASSUNG
The objective of this study was to look for optimal preparation of hydrolysates of desalted duck egg white powder (DDEWP) by the three different proteases and to investigate their antioxidant and antimicrobial properties. DDEWP was hydrolyzed by three proteases, including pepsin (PEP), Bacillus spp. (BA) and natokinase (NAT) with three different enzyme concentrations (0.1%, 0.3%, and 0.5%), individually. The important key hydrolysis parameters such as hydrolysis degree, yield, antioxidant and antimicrobial activity were evaluated in this experiment. The results showed that the degree of hydrolysis (DH) of all treatments increased with increasing hydrolysis time and protease concentrations. The antioxidant and antimicrobial activities of the hydrolysates were affected by type and concentration of protease as well as hydrolysis time. Hydrolysis of PEP significantly (p<0.05) obtained the highest yield of hydrolysates, however, both of BA and NAT showed substantially lower DH values and still did not exceed 5% by the end of hydrolysis. Among the different hydrolysates, PEP exhibited significantly higher 2, 2-diphenyl-1-picrylhydrazyl (DPPH) radical scavenging activity than BA and NAT. All DDEWP hydrolysates from PEP had low ferrous ion chelating activity (<37%) that was significantly lower than that of NAT (>37% to 92%) and BA (30% to 79%). Besides, DDEWP hydrolysates of PEP presented significantly higher reducing power than BA and NAT. In antimicrobial activities, Escherichia coli, Salmonella typhimurium, and Pseudomonas aeruginosa were not effectively inhibited by any DDEWP hydrolysates of PEP except for Staphylococcus aureus. Especially, the excellent antibacterial activity against S. aureus only was displayed in DDEWP hydrolysates of PEP 0.1%. DDEWP hydrolysates from PEP demonstrated significantly better DH, yield, DPPH radical scavenging activity and reducing power, furthermore, had excellent inhibitory on S. aureus due to large clear zone and moderated inhibitory in bactericidal inhibition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphoenolbrenztraubensäure Trinatriumsalz Hydrat, ≥97% (enzymatic)