- Flavonoid compounds are potent inhibitors of cyclic AMP phosphodiesterase.
Flavonoid compounds are potent inhibitors of cyclic AMP phosphodiesterase.
Experientia (1978-08-15)
A Beretz, R Anton, J C Stoclet
PMID212288
ZUSAMMENFASSUNG
The inhibitory activity of 19 flavonoid molecules on cyclic AMP breakdown by a commercial beef heart phosphodiesterase preparation is reported. 7 compounds are active in the micromolar range, 2 of which have a potency equivalent to that of papaverine. Some structure activity relationships are drawn.