Merck
  • Optimization of β-mannanase production by Bacillus subtilis US191 using economical agricultural substrates.
Optimization of β-mannanase production by Bacillus subtilis US191 using economical agricultural substrates.
Biotechnology progress (2020-03-07)
Monia Blibech, Ameny Farhat-Khemakhem, Mouna Kriaa, Rania Aslouj, Ines Boukhris, Othman A Alghamdi, Hichem Chouayekh
ZUSAMMENFASSUNG
The Bacillus subtilis US191 strain producing highly thermostable β-mannanase was previously selected as potential probiotic candidate for application as feed supplement in poultry industry. Initially, the level of extracellular β-mannanase production by this strain was 1.48 U ml-1 . To improve this enzyme titer, the present study was undertaken to optimize the fermentation conditions through experimental designs and valorization of agro-industrial byproducts. Using the Plackett-Burman design, in submerged fermentation, a set of 14 culture variables was evaluated in terms of their effects on β-mannanase production. Locust bean gum (LBG), soymeal, temperature, and inoculum size were subsequently optimized by response surface methodology using Box-Behnken design. Under optimized conditions (1 g L-1 LBG, 8 g L-1 soymeal, temperature of 30°C and inoculum size of 1010  CFU ml-1 ), a 2.59-fold enhancement in β-mannanase titer was achieved. Next, to decrease the enzyme production cost, the effect of partial substitution of LBG (1 g L-1 ) by agro-industrial byproducts was investigated, and a Taguchi design was applied. This allowed the attaining of a β-mannanase production level of 8.75 U ml-1 in presence of 0.25 g L-1 LBG, 5 g L-1 of coffee residue powder, 5 g L-1 of date seeds powder, and 5 g L-1 of prickly pear seeds powder as mannans sources. Overall, a 5.91-fold improvement in β-mannanase production by B. subtilis US191 was achieved.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Locust bean gum from Ceratonia siliqua seeds