Merck
  • Phase I and Pharmacokinetic Study of Romidepsin in Patients with Cancer and Hepatic Dysfunction: A National Cancer Institute Organ Dysfunction Working Group Study.
Phase I and Pharmacokinetic Study of Romidepsin in Patients with Cancer and Hepatic Dysfunction: A National Cancer Institute Organ Dysfunction Working Group Study.
Clinical cancer research : an official journal of the American Association for Cancer Research (2020-08-21)
Roisin M Connolly, Eric Laille, Ulka Vaishampayan, Vincent Chung, Karen Kelly, Afshin Dowlati, Olatunji B Alese, R Donald Harvey, Paul Haluska, Lillian L Siu, Shivaani Kummar, Richard Piekarz, S Percy Ivy, Nicole M Anders, Melinda Downs, Ashley O'Connor, Angela Scardina, Jacqueline Saunders, Gary L Rosner, Michael A Carducci, Michelle A Rudek
ZUSAMMENFASSUNG
Romidepsin dosing recommendations for patients with malignancy and varying degrees of hepatic dysfunction was lacking at the time of regulatory approval for T-cell lymphoma. We conducted a multicenter phase I clinical trial (ETCTN-9008) via the NCI Organ Dysfunction Working Group to investigate safety, first cycle MTD, and pharmacokinetic profile of romidepsin in this setting. Patients with select advanced solid tumors or hematologic malignancies were stratified according to hepatic function. Romidepsin was administered intravenously on days 1, 8, and 15 of a 28-day cycle and escalation followed a 3 + 3 design in moderate and severe impairment cohorts. Blood samples for detailed pharmacokinetic analyses were collected after the first dose. Thirty-one patients received one dose of romidepsin and were evaluable for pharmacokinetic analyses in normal (n = 12), mild (n = 8), moderate (n = 5), and severe (n = 6) cohorts. Adverse events across cohorts were similar, and dose-limiting toxicity occurred in two patients (mild and severe impairment cohorts). The MTD was not determined because the geometric mean AUC values of romidepsin in moderate (7 mg/m2) and severe (5 mg/m2) impairment cohort were 114% and 116% of the normal cohort (14 mg/m2). Data from the ETCTN-9008 trial led to changes in the romidepsin labeling to reflect starting dose adjustment for patients with cancer and moderate and severe hepatic impairment, with no adjustment for mild hepatic impairment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Romidepsin, ≥98% (HPLC)