Merck
  • Members of the miR-30 family inhibit the epithelial-to-mesenchymal transition of non-small-cell lung cancer cells by suppressing XB130 expression levels.
Members of the miR-30 family inhibit the epithelial-to-mesenchymal transition of non-small-cell lung cancer cells by suppressing XB130 expression levels.
Oncology letters (2020-08-31)
Kewei Song, Yinhui Jiang, Yan Zhao, Yuan Xie, Jianjiang Zhou, Wenfeng Yu, Qinrong Wang
ZUSAMMENFASSUNG
MicroRNAs (miRs) are associated with cancer metastasis. Aberrant expression levels of members of the miR-30 family have been observed in non-small-cell lung cancer (NSCLC). However, the effects of miR-30 family members on the epithelial-to-mesenchymal transition (EMT) of NSCLC cells and the underlying molecular mechanisms have not yet been fully elucidated. The present study investigated the effects of miR-30 family members on EMT, migration and invasion of NSCLC cells and found that overexpression of these miRs inhibited EMT via decreasing the expression levels of N-cadherin, β-catenin and SNAI1, along with weakened migration and invasion abilities. Then, XB130 was identified as a downstream target of the miR-30 family members. XB130-knockdown also inhibited EMT of NSCLC cells, whereas ectopic overexpression of XB130 partly rescued the suppressive effects of miR-30c and miR-30d on EMT. In conclusion, miR-30 family members inhibited EMT of NSCLC cells, partially via suppressing XB130 expression levels.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human AFAP1L2