Merck
  • An outbreak of cutaneous abscesses caused by Panton-Valentine leukocidin-producing methicillin-susceptible Staphylococcus aureus among gold mine workers, South Africa, November 2017 to March 2018.
An outbreak of cutaneous abscesses caused by Panton-Valentine leukocidin-producing methicillin-susceptible Staphylococcus aureus among gold mine workers, South Africa, November 2017 to March 2018.
BMC infectious diseases (2020-08-25)
Husna Ismail, Nelesh P Govender, Ashika Singh-Moodley, Erika van Schalkwyk, Liliwe Shuping, Itumeleng Moema, Gal Feller, Ruth Mogokotleng, Wilhelmina Strasheim, Michelle Lowe, Ruth Mpembe, Serisha Naicker, Tsidiso G Maphanga, Cecilia De Abreu, Farzana Ismail, Nazir Ismail, Mushal Allam, Arshad Ismail, Tanusha Singh, Onnicah Matuka, Thabang Duba, Olga Perovic
ZUSAMMENFASSUNG
We aimed to describe an outbreak of cutaneous abscesses caused by Panton-Valentine leukocidin (PVL)-producing methicillin-susceptible Staphylococcus aureus (MSSA) among gold mine workers. In February 2018, we retrospectively reviewed a random sample of 50 medical records from 243 cases and conducted face-to-face interviews using a structured questionnaire. Pus aspirates were sent to the National Institute for Communicable Diseases from prospectively-identified cases (November 2017-March 2018). Nasopharyngeal swabs were collected during a colonisation survey in February 2018. Staphylococcus aureus isolates were screened with a conventional PCR for lukS/F-PV. Pulsed-field gel electrophoresis (PFGE) was performed to determine the genetic relatedness among the isolates. A sample of isolates were selected for whole genome sequencing (WGS). We conducted an assessment on biological risks associated with mining activities. From January 2017 to February 2018, 10% (350/3582) of mine workers sought care for cutaneous abscesses. Forty-seven medical files were available for review, 96% were male (n = 45) with a mean age of 43 years (SD = 7). About 52% (24/46) were involved in stoping and 28% (13/47) worked on a particular level. We cultured S. aureus from 79% (30/38) of cases with a submitted specimen and 14% (12/83) from colonisation swabs. All isolates were susceptible to cloxacillin. Seventy-one percent of S. aureus isolates (30/42) were PVL-PCR-positive. Six PFGE clusters were identified, 57% (21/37) were closely related. WGS analysis found nine different sequence types. PFGE and WGS analysis showed more than one cluster of S. aureus infections involving closely related isolates. Test reports for feed and product water of the mine showed that total plate counts were above the limits of 1000 cfu/ml, coliform counts > 10 cfu/100 ml and presence of faecal coliforms. Best practices were poorly implemented as some mine workers washed protective clothing with untreated water and hung them for drying at the underground surface. PVL-producing MSSA caused an outbreak of cutaneous abscesses among underground workers at a gold mining company. To our knowledge, no other outbreaks of PVL-producing S. aureus involving skin and soft tissue infections have been reported in mining facilities in South Africa. We recommend that worker awareness of infection prevention and control practices be strengthened.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dess-Martin-Periodinan, 97%