Merck
  • Vapor-assisted deposition of highly efficient, stable black-phase FAPbI3 perovskite solar cells.
Vapor-assisted deposition of highly efficient, stable black-phase FAPbI3 perovskite solar cells.
Science (New York, N.Y.) (2020-10-03)
Haizhou Lu, Yuhang Liu, Paramvir Ahlawat, Aditya Mishra, Wolfgang R Tress, Felix T Eickemeyer, Yingguo Yang, Fan Fu, Zaiwei Wang, Claudia E Avalos, Brian I Carlsen, Anand Agarwalla, Xin Zhang, Xiaoguo Li, Yiqiang Zhan, Shaik M Zakeeruddin, Lyndon Emsley, Ursula Rothlisberger, Lirong Zheng, Anders Hagfeldt, Michael Grätzel
ZUSAMMENFASSUNG
Mixtures of cations or halides with FAPbI3 (where FA is formamidinium) lead to high efficiency in perovskite solar cells (PSCs) but also to blue-shifted absorption and long-term stability issues caused by loss of volatile methylammonium (MA) and phase segregation. We report a deposition method using MA thiocyanate (MASCN) or FASCN vapor treatment to convert yellow δ-FAPbI3 perovskite films to the desired pure α-phase. NMR quantifies MA incorporation into the framework. Molecular dynamics simulations show that SCN- anions promote the formation and stabilization of α-FAPbI3 below the thermodynamic phase-transition temperature. We used these low-defect-density α-FAPbI3 films to make PSCs with >23% power-conversion efficiency and long-term operational and thermal stability, as well as a low (330 millivolts) open-circuit voltage loss and a low (0.75 volt) turn-on voltage of electroluminescence.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-tert-Butylpyridin, 98%