Merck
  • Farnesoid X receptor and liver X receptors regulate Oct3/4 expression by multiple feedback regulating system in normal renal-derived cells and renal adenocarcinoma cells.
Farnesoid X receptor and liver X receptors regulate Oct3/4 expression by multiple feedback regulating system in normal renal-derived cells and renal adenocarcinoma cells.
The Journal of toxicological sciences (2020-01-15)
Tomofumi Fujino, Kouta Sugizaki, Rei Kato, Masaki Beppu, Satoshi Murakami, Hyunjung Lee, Toshiyuki Oshima, Makio Hayakawa
ZUSAMMENFASSUNG
In this study, we found that nuclear receptors FXR and LXR (originally characterized as regulatory factors involved in cholesterol/bile acid homeostasis) regulate the expression of Oct3/4, a marker for cell differentiation, in both normal renal-derived cell line HK-2 and renal adenocarcinoma cell line ACHN. Down-regulation of Oct3/4 expression by activating FXR and LXR occurs only in normal renal cell-derived HK-2 cells. We also found that the RNA-binding protein, ELAVL2, oppositely regulates Oct3/4 expressions in HK-2 and ACHN cells. Moreover, we revealed that LXR-alpha and LXR-beta regulate each other's expression. Although an LXR-beta-specific agonist is assumed to be the basis for an anti-arteriosclerotic drug that only stimulates reverse cholesterol transport, our findings show that the development of such an anti-arteriosclerotic drug would require further elucidation of the complex mechanism of LXR-alpha and LXR-beta regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%
Sigma-Aldrich
GW4064, ≥97% (HPLC)