Merck
  • Extravascular transport of drugs in tumor tissue: effect of lipophilicity on diffusion of tirapazamine analogues in multicellular layer cultures.
Extravascular transport of drugs in tumor tissue: effect of lipophilicity on diffusion of tirapazamine analogues in multicellular layer cultures.
Journal of medicinal chemistry (2005-02-18)
Frederik B Pruijn, Joanna R Sturman, H D Sarath Liyanage, Kevin O Hicks, Michael P Hay, William R Wilson
ZUSAMMENFASSUNG
The extravascular diffusion of antitumor agents is a key determinant of their therapeutic activity, but the relationships between physicochemical properties of drugs and their extravascular transport are poorly understood. It is well-known that drug lipophilicity plays an important role in transport across biological membranes, but the net effect of lipophilicity on transport through multiple layers of tumor cells is less clear. This study examines the influence of lipophilicity (measured as the octanol-water partition coefficient P) on the extravascular transport properties of the hypoxic cytotoxin tirapazamine (TPZ, 1) and a series of 13 neutral analogues, using multicellular layers (MCLs) of HT29 human colon carcinoma cells as an in vitro model for the extravascular compartment of tumors. Flux of drugs across MCLs was determined using diffusion chambers, with the concentration-time profile on both sides of the MCL measured by HPLC. Diffusion coefficients in the MCLs (D(MCL)) were inversely proportional to M(r)(0.5) (M(r), relative molecular weight), although this was a minor contributor to differences between compounds over the narrow M(r) range investigated. Differences in lipophilicity had a larger effect, with a sigmoidal dependence of D(MCL) on log P. Correcting for M(r) differences, lipophilic compounds (log P > 1.5) had ca. 15-fold higher D(MCL) than hydrophilic compounds (log P < -1). Using a pharmacokinetic/pharmacodynamic (PK/PD) model in which diffusion in the extravascular compartment of tumors is considered explicitly, we demonstrated that hypoxic cell kill is very sensitive to changes in extravascular diffusion coefficient of TPZ analogues within this range. This study shows that simple monosubstitution of TPZ can alter log P enough to markedly improve extravascular transport and activity against target cells, especially if rates of metabolic activation are also optimized.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
9(10H)-Acridanon, 99%
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination