Merck
  • High Contrast Surface Enhanced Fluorescence of Carbon Dot Labeled Bacteria Cells on Aluminum Foil.
High Contrast Surface Enhanced Fluorescence of Carbon Dot Labeled Bacteria Cells on Aluminum Foil.
Journal of fluorescence (2020-08-29)
Rostislav Bukasov, Zhanar Kunushpayeva, Alisher Rapikov, Saida Zhunussova, Alisher Sultangaziyev, Olena Filchakova
ZUSAMMENFASSUNG
Surface enhanced fluorescence (SEF) is observed with very high contrast (100-200) from single E. coli bacteria cells labeled with Carbon nanodots (CDs), on aluminum foil and aluminum film. Likely, it is the first application of organic CDs in SEF. SEF with 633 nm excitation delivered a much higher contrast than SEF with 532 nm excitation. Contrast is the ratio of the fluorescent intensities of labeled CDs to unlabeled (control) cells. High contrast with CDs is also observed on the gold film, silicon, and glass. Enhancement factor (EF) is the ratio of the signal on the metal substrate to the signal on the glass. Single E. coli cells, labeled with commercial graphene quantum dots (GCDs), demonstrated higher EFs (44 on gold, 35 on Al film), but at least one order of magnitude lower contrast (7-10 on aluminum and gold) than cells labeled with organic CDs. Therefore, organic CDs can be a good choice for cell imaging/labeling, capable of achieving a signal to noise (standard deviation of the control) as high as 700 on Al film. Overall, aluminum foil and film are highlighted as inexpensive but efficient substrates for Metal Enhanced Fluorescence, particularly MEF of bacterial cells stained with CDs.