Merck
  • RAB25 confers resistance to chemotherapy by altering mitochondrial apoptosis signaling in ovarian cancer cells.
RAB25 confers resistance to chemotherapy by altering mitochondrial apoptosis signaling in ovarian cancer cells.
Apoptosis : an international journal on programmed cell death (2020-09-10)
Sehime Gulsun Temel, Aslı Giray, Bahriye Karakas, Ozgur Gul, Ilknur Kozanoglu, Husnu Celik, Huveyda Basaga, Ufuk Acikbas, Ceren Sucularli, Sidika Oztop, Yeliz Aka, Ozgur Kutuk
ZUSAMMENFASSUNG
Ovarian cancer remains one of the most frequent causes of cancer-related death in women. Many patients with ovarian cancer suffer from de novo or acquired resistance to chemotherapy. Here, we report that RAB25 suppresses chemotherapy-induced mitochondrial apoptosis signaling in ovarian cancer cell lines and primary ovarian cancer cells. RAB25 blocks chemotherapy-induced apoptosis upstream of mitochondrial outer membrane permeabilization by either increasing antiapoptotic BCL-2 proteins or decreasing proapoptotic BCL-2 proteins. In particular, BAX expression negatively correlates with RAB25 expression in ovarian cancer cells. BH3 profiling assays corroborated that RAB25 decreases mitochondrial cell death priming. Suppressing RAB25 by means of RNAi or RFP14 inhibitory hydrocarbon-stapled peptide sensitizes ovarian cancer cells to chemotherapy as well as RAB25-mediated proliferation, invasion and migration. Our data suggest that RAB25 is a potential therapeutic target for ovarian cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%
Sigma-Aldrich
MISSION® esiRNA, targeting human RAB25