Merck
Influence of Surface Conditioning on the Repair Strength of Bioactive Restorative Material.
Journal of applied biomaterials & functional materials (2020-09-18)
Mohamed Moustafa Awad, Nader Almutairi, Feras Alhalabi, Ali Robaian, Fahim Ahmed Vohra, Mutlu Ozcan, Ahmed Maawadh, Ali Alrahlah
ZUSAMMENFASSUNG
To evaluate the effects of surface treatment and repair material on the repair shear bond strength (SBS) of the bioactive restorative material. A total of 240 Activa BioActive Restorative (Activa) discs were prepared, aged, and polished, and divided randomly into eight groups (n = 30). Groups 1-4 discs were repaired with bulk-fill flowable resin-based composite (Bulk-RBC), and Groups 5-8 discs with Activa. Surface treatment used for each repair material type were air abrasion with silica-coated 30-m Al2O3 particles (air abrasion) (Groups 2 & 6), Air abrasion with universal primer (Groups 3 & 7), and Air abrasion with universal adhesive (Groups 4 & 8). Groups 1 and 5 were controls without surface treatment. SBS test was performed, and the failure mode and surface topography were assessed. Surface treatment with air abrasion significantly improved the SBS for repair using both Activa and Bulk-RBC. Repair SBS using Activa was significantly higher compared with Bulk-RBC. Cohesive failure in substrate and mixed failures were most common in the surface-treated groups (2-4, 6-8). Air abrasion produced prominent surface topography changes compared with polishing. Air abrasion enhances the repair SBS of aged bioactive restorative material. The use of the same material (Activa) for repair affords a higher bond strength compared with the use bulk-RBC.