Merck
Novel and potent antimicrobial effects of caspofungin on drug-resistant Candida and bacteria.
Scientific reports (2020-10-22)
Makoto Sumiyoshi, Taiga Miyazaki, Juliann Nzembi Makau, Satoshi Mizuta, Yoshimasa Tanaka, Takeshi Ishikawa, Koichi Makimura, Tatsuro Hirayama, Takahiro Takazono, Tomomi Saijo, Hiroyuki Yamaguchi, Shintaro Shimamura, Kazuko Yamamoto, Yoshifumi Imamura, Noriho Sakamoto, Yasushi Obase, Koichi Izumikawa, Katsunori Yanagihara, Shigeru Kohno, Hiroshi Mukae
ZUSAMMENFASSUNG
Echinocandins, including caspofungin, micafungin, and anidulafungin, are first-line antifungal agents for the treatment of invasive candidiasis. They exhibit fungicidal activity by inhibiting the synthesis of β-1,3-D-glucan, an essential component of the fungal cell wall. However, they are active only against proliferating fungal cells and unable to completely eradicate fungal cells even after a 24 h drug exposure in standard time-kill assays. Surprisingly, we found that caspofungin, when dissolved in low ionic solutions, had rapid and potent antimicrobial activities against multidrug-resistant (MDR) Candida and bacteria cells even in non-growth conditions. This effect was not observed in 0.9% NaCl or other ion-containing solutions and was not exerted by other echinocandins. Furthermore, caspofungin dissolved in low ionic solutions drastically reduced mature biofilm cells of MDR Candida auris in only 5 min, as well as Candida-bacterial polymicrobial biofilms in a catheter-lock therapy model. Caspofungin displayed ion concentration-dependent conformational changes and intracellular accumulation with increased reactive oxygen species production, indicating a novel mechanism of action in low ionic conditions. Importantly, caspofungin dissolved in 5% glucose water did not exhibit increased toxicity to human cells. This study facilitates the development of new therapeutic strategies in the management of catheter-related biofilm infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anidulafungin, ≥97% (HPLC)