Merck
Biology of the RANTES/SIS cytokine family.
Cytokine (1991-05-01)
T J Schall
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MIP-1 beta (CCL4) human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
RANTES human, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
MIP-1 beta from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture