Merck
  • Solid-Liquid Europium Ion Extraction via Phosphonic Acid-Functionalized Polyvinylidene Fluoride Siloxanes.
Solid-Liquid Europium Ion Extraction via Phosphonic Acid-Functionalized Polyvinylidene Fluoride Siloxanes.
Polymers (2020-09-03)
Mohammad Wehbi, Ahmad Mehdi, Ali Alaaeddine, Nada Jaber, Bruno Ameduri
ZUSAMMENFASSUNG
Novel triethoxysilane and dimethyl phosphonate functional vinylidene fluoride (VDF)-containing terpolymers, for potential applications in Eu ion extraction from water, were produced by conventional radical terpolymerization of VDF with vinyltriethoxylsilane (VTEOS) and vinyldimethylphosphonate (VDMP). Although initial attempts for the copolymerization of VTEOS and VDMP failed, the successful terpolymerization was initiated by peroxide to lead to multiple poly(VDF-ter-VDMP-ter-VTEOS) terpolymers, that had different molar percentages of VDF (70-90 mol.%), VTEOS (5-20 mol.%) and VDMP (10 mol.%) in 50-80% yields. The obtained terpolymers were characterized by 1H, 19F, 29Si and 31P NMR spectroscopies. The crosslinking of such resulting poly(VDF-ter-VDMP-ter-VTEOS) terpolymers was achieved by hydrolysis and condensation (sol-gel process) of the triethoxysilane groups in acidic media, to obtain a 3D network, which was analyzed by solid state 29Si and 31P NMR spectroscopies, TGA and DSC. The thermal stability of the terpolymers was moderately high (up to 300 °C under air), whereas they display a slight increase in their crystallinity-rate from 9.7% to 12.1% after crosslinking. Finally, the dimethyl phosphonate functions were hydrolyzed into phosphonic acid successfully, and the europium ion extraction capacity of terpolymer was studied. The results demonstrated a very high removal capacity of Eu(III) ions from water, up to a total removal at low concentrations.