Merck
  • A new synthesis of isoaurones: cytotoxic activity of compounds related to the alleged structure of isoaurostatin.
A new synthesis of isoaurones: cytotoxic activity of compounds related to the alleged structure of isoaurostatin.
Bioorganic & medicinal chemistry letters (2005-07-26)
Eleonora Rizzi, Sabrina Dallavalle, Lucio Merlini, Giovanni Luca Beretta, Graziella Pratesi, Franco Zunino
ZUSAMMENFASSUNG
A new synthesis of isoaurones related to the alleged structure of isoaurostatin, via Heck intramolecular cyclization of cinnamic esters of 2-iodophenols, is reported. The cytotoxic activity of these isoaurones is lower than that of the structurally very similar 4-arylcoumarins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Daidzein, ≥98%, synthetic