Merck
  • Preparation and characterization of microparticles of ?-cyclodextrin/glutathione and chitosan/glutathione obtained by spray-drying
Preparation and characterization of microparticles of ?-cyclodextrin/glutathione and chitosan/glutathione obtained by spray-drying
Food Research International (2018)
Webber V, et al.