Merck
  • Presentations of stroke and acute myocardial infarction in the first 28 days following the introduction of State of Emergency restrictions for COVID-19.
Presentations of stroke and acute myocardial infarction in the first 28 days following the introduction of State of Emergency restrictions for COVID-19.
Emergency medicine Australasia : EMA (2020-08-25)
Biswadev Mitra, Rob D Mitchell, Geoffrey C Cloud, Dion Stub, Minh Nguyen, Shane Nanayakkara, Jean-Philippe Miller, Gerard M O'Reilly, De Villiers Smit, Peter A Cameron
ZUSAMMENFASSUNG
To determine if Victorian State of Emergency (SOE) measures to combat COVID-19 were associated with delayed presentations or management of acute stroke and acute myocardial infarction (AMI). This was a retrospective, pre- and post-implementation study using data from an adult, tertiary cardiology and neurosciences centre with 24-h capacity for endovascular procedures. All primary presentations with acute stroke or AMI during the first 28 days of stage 2 and stage 3 SOE restrictions (26 March to 23 April 2020) were compared to an equivalent period without restrictions (26 March to 23 April 2019). The primary outcome variable was time from onset of symptoms to ED presentation. There were 52 (1.6% of all ED presentations) patients who met inclusion criteria during the SOE period and 57 (1.0%) patients in the comparator period. Patients were equally matched for demographics, disease severity and prior history of stroke or AMI. Median time from symptom onset to presentation was 227 (93-1183) min during the SOE period and 342 (119-1220) min during the comparator period (P = 0.24). Among eligible patients with ischaemic stroke or ST-elevation AMI, median time to primary reperfusion intervention was 65 (37-78) min during SOE and 44 (39-60) min in the comparator period (P = 0.54). There were no differences in mortality at hospital discharge (9.6% vs 10.5%) and hospital length of stay (5.4 vs 4.3 days). In the first 28 days, SOE measures to combat COVID-19 were not associated with delays in presentation or life-saving interventions for patients with acute stroke and AMI.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
CTB, ≥98% (HPLC)