Merck
  • Pathogen flows from on-site sanitation systems in low-income urban neighborhoods, Dhaka: A quantitative environmental assessment.
Pathogen flows from on-site sanitation systems in low-income urban neighborhoods, Dhaka: A quantitative environmental assessment.
International journal of hygiene and environmental health (2020-09-18)
Nuhu Amin, Pengbo Liu, Tim Foster, Mahbubur Rahman, Md Rana Miah, Golam Bashir Ahmed, Mamun Kabir, Suraja Raj, Christine L Moe, Juliet Willetts
ZUSAMMENFASSUNG
Despite wide usage of on-site sanitation, there is limited field-based evidence on the removal or release of pathogens from septic tanks and other primary treatment systems, such as anaerobic baffled reactors (ABR). In two low-income areas in Dhaka, we conducted a cross-sectional study to explore pathogen loads discharged from commonly used on-site sanitation-systems and their transport in nearby drains and waterways. We collected samples of drain water, drain sediment, canal water, and floodwater from April-October 2019. Sludge, supernatant, and effluent samples were also collected from septic tanks and ABRs. We investigated the presence and concentration of selected enteric pathogens (Shigella, Vibrio cholerae (V. cholerae), Salmonella Typhi (S. Typhi), Norovirus Genogroup-II (NoV-GII), and Giardia) and presence of Cryptosporidium in these samples using quantitative polymerase chain reaction (qPCR).The equivalent genome copies (EGC) of individual pathogens were estimated in each sample by interpolation of the mean Ct value to the corresponding standard curve and the dilution factor for each sample type. Absolute quantification was expressed as log10 EGC per 100 mL for the water samples and log10 EGC per gram for the sediment samples. Among all samples tested (N = 151), 89% were contaminated with Shigella, 68% with V. cholerae and NoV-GII, 32% with Giardia, 17% with S. Typhi and 6% with Cryptosporidium. A wide range of concentration of pathogens [range: mean log10 concentration of Giardia = 0.74 EGC/100 mL in drain ultrafiltration samples to mean log10 concentration of NoV-GII and Giardia = 7.11 EGC/100 mL in ABR sludge] was found in all environmental samples. The highest pathogen concentrations were detected in open drains [range: mean log10 concentration = 2.50-4.94 EGC/100 mL], septic tank effluent [range: mean log10 concentration = 3.32-4.65 EGC/100 mL], and ABR effluent [range: mean log10 concentration = 2.72-5.13 EGC/100 mL]. High concentrations of pathogens (particularly NoV-GII, V.cholerae and Shigella) were frequently detected in environmental samples from two low-income urban neighbourhoods of Dhaka city. The numerous environmental exposure pathways for children and adults make these findings of public health concern. These results should prompt rethinking of how to achieve safe sanitation solutions that protect public health in dense low-income areas. In particular, improved management and maintenance regimes, further treatment of liquid effluent from primary treatment processes, and appropriate application of onsite, decentralised and offsite sanitation systems given the local context.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Albumin aus Rinderserum, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Glas-Beads, säuregereinigt, 212-300 μm (50-70 U.S. sieve)