Merck
  • New NO-releasing pharmacodynamic hybrids of losartan and its active metabolite: design, synthesis, and biopharmacological properties.
New NO-releasing pharmacodynamic hybrids of losartan and its active metabolite: design, synthesis, and biopharmacological properties.
Journal of medicinal chemistry (2006-04-14)
Maria C Breschi, Vincenzo Calderone, Maria Digiacomo, Marco Macchia, Alma Martelli, Enrica Martinotti, Filippo Minutolo, Simona Rapposelli, Armando Rossello, Lara Testai, Aldo Balsamo
ZUSAMMENFASSUNG
In a preliminary work, we reported two NO-sartans, possessing the characteristics of an AT(1) antagonist and a "slow NO donor", obtained by adding NO-donor side chains to losartan 1. The NO release from an NO-sartan should be modulated in order to strengthen the antihypertensive activity of the native drug and to ensure additional effects, such as the antiplatelet and anti-ischemic ones. To obtain a collection of prototypical NO-sartans, showing different rates of NO release, new NO-donor moieties have been linked to 1 or its active metabolite 2 (EXP 3174). Almost all the synthesized compounds exhibited both AT(1)-antagonist and NO-mediated vasorelaxing properties, with a wide range of NO-releasing rates. Further pharmacological investigation on compound 4a showed that it possessed antihypertensive and cardiac antihypertrophic effects similar to those of the reference AT(1)-blocking or ACE-inhibiting drugs. Furthermore, the additional anti-ischemic cardio-protective properties and antiplatelet effects of 4a have been preliminarily investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Captopril, ≥98% (HPLC), powder
Sigma-Aldrich
Captopril, meets USP testing specifications