Merck
  • Expression of DNA Methyltransferase 1 Is a Hallmark of Melanoma, Correlating with Proliferation and Response to B-Raf and Mitogen-Activated Protein Kinase Inhibition in Melanocytic Tumors.
Expression of DNA Methyltransferase 1 Is a Hallmark of Melanoma, Correlating with Proliferation and Response to B-Raf and Mitogen-Activated Protein Kinase Inhibition in Melanocytic Tumors.
The American journal of pathology (2020-07-18)
Maximilian Gassenmaier, Maximilian Rentschler, Birgit Fehrenbacher, Thomas K Eigentler, Kristian Ikenberg, Corinna Kosnopfel, Tobias Sinnberg, Heike Niessner, Hans Bösmüller, Nikolaus B Wagner, Martin Schaller, Claus Garbe, Martin Röcken
ZUSAMMENFASSUNG
Aberrant DNA methylation is an epigenetic hallmark of melanoma, but the expression of DNA methyltransferase (Dnmt)-1 in melanocytic tumors is unknown. Dnmt1 expression was analyzed in primary melanocytes, melanoma cell lines, and 83 melanocytic tumors, and its associations with proliferation, mutational status, and response to B-Raf and mitogen-activated protein kinase kinase (MEK) inhibition were explored. Dnmt1 expression was increased incrementally from nevi [mean fluorescence intensity (MFI), 48.1; interquartile range, 41.7 to 59.6] to primary melanomas (MFI, 68.8; interquartile range, 58.4 to 77.0) and metastatic melanomas (MFI, 87.5; interquartile range, 77.1 to 114.5) (P < 0.001). Dnmt1 expression was correlated with Ki-67 expression (Spearman correlation, 0.483; P < 0.001) and was independent of BRAF mutation status (P = 0.55). In BRAF-mutant melanoma, Dnmt1 was down-regulated during response to B-Raf and MEK inhibition and was again up-regulated on drug resistance in vitro and in vivo. Degradation of Dnmt1 by the histone deacetylase inhibitor suberoylanilide hydroxamic acid was associated with decreased cell viability in B-Raf inhibitor-sensitive and -resistant cell lines. This study demonstrates that Dnmt1 expression is correlated with proliferation in melanocytic tumors, is increased with melanoma progression, and is associated with response to B-Raf and MEK inhibition. Given its strong expression in metastatic melanoma, Dnmt1 may be a promising target for combined epigenetic and immunotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
4-Methylumbelliferyl-Heptanoat, ≥95% (GC)