Merck
Controlling Particle Morphology and Pore Size in the Synthesis of Ordered Mesoporous Materials.
Molecules (Basel, Switzerland) (2020-10-30)
Yaregal Awoke, Yonas Chebude, Isabel Díaz
ZUSAMMENFASSUNG
Ordered mesoporous materials have attracted considerable attention due to their potential applications in catalysis, adsorption, and separation technologies, as well as biomedical applications. In the present manuscript, we aim at a rational design to obtain the desired surface functionality (Ti and/or hydrophobic groups) while obtaining short channels (short diffusion paths) and large pore size (>10 nm). Santa Barbara Amorphous material SBA-15 and periodic mesoporous organosilica PMO materials are synthesized using Pluronic PE 10400 (P104) surfactant under mild acidic conditions to obtain hexagonal platelet-like particles with very short mesochannels (300-450 nm). The use of expanders, such as 1, 3, 5-trimethylbenzene (TMB) and 1, 3, 5-triisopropylbenzene (TIPB) were tested in order to increase the pore size. TMB yielded in the formation of vesicles in all the syntheses attempted, whereas P104 combined with TIPB resulted both in expanded (E) E-SBA-15 and E-PMO with 12.3 nm pore size short channel particles in both cases. Furthermore, the synthesis method was expanded to the incorporation of small amount of Ti via co-condensation method using titanocene as titanium source. As a result, Ti-E-SBA-15 was obtained with 15.5 nm pore size and isolated Ti-sites maintaining platelet hexagonal morphology. Ti-PMO was obtained with 7.8 nm and short channels, although the pore size under the tried synthesis conditions could not be expanded further without losing the structural ordering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Bis(triethoxysilyl)ethan, 96%