Merck
  • Synapse type-specific proteomic dissection identifies IgSF8 as a hippocampal CA3 microcircuit organizer.
Synapse type-specific proteomic dissection identifies IgSF8 as a hippocampal CA3 microcircuit organizer.
Nature communications (2020-10-16)
Nuno Apóstolo, Samuel N Smukowski, Jeroen Vanderlinden, Giuseppe Condomitti, Vasily Rybakin, Jolijn Ten Bos, Laura Trobiani, Sybren Portegies, Kristel M Vennekens, Natalia V Gounko, Davide Comoletti, Keimpe D Wierda, Jeffrey N Savas, Joris de Wit
ZUSAMMENFASSUNG
Excitatory and inhibitory neurons are connected into microcircuits that generate circuit output. Central in the hippocampal CA3 microcircuit is the mossy fiber (MF) synapse, which provides powerful direct excitatory input and indirect feedforward inhibition to CA3 pyramidal neurons. Here, we dissect its cell-surface protein (CSP) composition to discover novel regulators of MF synaptic connectivity. Proteomic profiling of isolated MF synaptosomes uncovers a rich CSP composition, including many CSPs without synaptic function and several that are uncharacterized. Cell-surface interactome screening identifies IgSF8 as a neuronal receptor enriched in the MF pathway. Presynaptic Igsf8 deletion impairs MF synaptic architecture and robustly decreases the density of bouton filopodia that provide feedforward inhibition. Consequently, IgSF8 loss impairs excitation/inhibition balance and increases excitability of CA3 pyramidal neurons. Our results provide insight into the CSP landscape and interactome of a specific excitatory synapse and reveal IgSF8 as a critical regulator of CA3 microcircuit connectivity and function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-vesikulärer Glutamattransporter 1-Antikörper, serum, Chemicon®
Sigma-Aldrich
Monoklonaler Anti-Synaptophysin-Antikörper in Maus hergestellte Antikörper, clone SVP-38, ascites fluid
Sigma-Aldrich
Mix-n-Stain CF 488A-Antikörpermarkierungskit (50–100μg)
Sigma-Aldrich
Anti-Polysialinsäure-NCAM-Antikörper, Klon 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-metabotroper Glutamatrezeptor 2/3-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Anti-BRINP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody