Merck
  • QSAR study on topically acting sulfonamides incorporating GABA moieties: a molecular connectivity approach.
QSAR study on topically acting sulfonamides incorporating GABA moieties: a molecular connectivity approach.
Bioorganic & medicinal chemistry letters (2006-02-07)
Vijay K Agrawal, Jyoti Singh, Padmakar V Khadikar, Claudiu T Supuran
ZUSAMMENFASSUNG
A quantitative Structure-activity relationship study (QSAR) on a set of carbonic anhydrase (CA, EC 4.2.1.1) inhibitors is reported using first-order valence connectivity index ((1)chi(v)). The inhibitory activity against three isozymes CAI, CAII (cystolic forms), and CAIV (membrane bound form), some of which are involved in important physiological processes, were considered for this purpose. All the three activities were excellently modeled by (1)chi(v) in multi-parametric regression containing indicator parameters. The results are critically discussed on the basis of various regression parameters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Sulfanilamid, ≥98%
Sigma-Aldrich
Sulfanilamid, puriss. p.a., ≥98% (calc. to the dried substance)
Supelco
Sulfanilamid, VETRANAL®, analytical standard