Merck
Analogues of N-hydroxycinnamoylphenalkylamides as inhibitors of human melanocyte-tyrosinase.
Bioorganic & medicinal chemistry letters (2006-01-31)
Sabrina Okombi, Delphine Rival, Sébastien Bonnet, Anne-Marie Mariotte, Eric Perrier, Ahcène Boumendjel
ZUSAMMENFASSUNG
Melanin play a major role in human skin protection and their biosynthesis is vital. Due to their color, they contribute to the skin pigmentation. Tyrosinase is a key enzyme involved in the first stage of melanin synthesis, catalyzing the transformation of tyrosine to l-dopaquinone. The aim of the present study was to study molecules able to inhibit melanin synthesis through inhibition of tyrosinase and their potential use in treating pigmentation-related disorders. We targeted amides obtained from coupling p-hydroxycinnamic acid derivatives with phenylalkylamines. The biological activity was evaluated on human melanocytes by an assay which measures tyrosine-catalyzed L-Dopa oxidation. The most active amides were: trans-N-caffeoyltyramine, N-dihydrocaffeoyltyramine, and trans-N-dihydro-p-hydroxycinnamoyltyramine which induce complete inhibition at 0.1mM. At the latter concentration, kojic acid, which was used as the reference inhibitor, was inactive.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
Kojisäure
Sigma-Aldrich
Tyramin, ≥98.0%
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Supelco
trans-Ferulasäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
trans-Ferulasäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland