Merck
  • Estimated 24-hour urinary sodium excretion as a risk factor for oxidative stress in Zambian adults: A cross-sectional study.
Estimated 24-hour urinary sodium excretion as a risk factor for oxidative stress in Zambian adults: A cross-sectional study.
PloS one (2020-11-13)
Violet Kayamba, Paul Kelly
ZUSAMMENFASSUNG
Persistent oxidative stress predisposes to various non-communicable diseases (NCDs), whose occurrence is increasing in sub-Saharan Africa. The aim of this study was to evaluate the link between markers of oxidative stress and some risk factors for NCDs in a Zambian cohort. We assessed oxidative stress by measuring 8-isoprostane (lipid oxidative stress) and 8-hydroxydeoxyguanosine (DNA oxidative stress). In addition, we measured mycotoxins (aflatoxin M1 and ochratoxin A), salt intake estimated from 24-hour sodium excretion calculated using the Tanaka and Kawaski formulae, and 1-hydroxypyrene (a metabolite of polycyclic aromatic hydrocarbons). Data on lifestyle risk factors were collected using questionnaires. Included were 244 participants; 128 (52%) were female and the median age was 48 years (IQR 39-58). The median level of 8-isoprostane was 0.13 ng/mg creatinine (IQR 0.08-0.23) while that of 8-hydroxydeoxyguanosine (8-OHdG) was 4 ng/mg creatinine (IQR 2-10). The median 24-hour sodium excretion was 21 g (IQR 16-25 g), with none being less than the 5 g recommended by WHO. Unadjusted urinary levels of 8-isoprostane were moderately correlated with 1-hydroxypyrene (Spearman r = 0.30, p<0.001) and estimated 24-hour urine sodium (Spearman r = 0.38, p<0.001). Urinary levels of 8-OHdG were not correlated with 1-hydroxypyrene, estimated 24-hour urine sodium, aflatoxin M1 or ochratoxin A (all p-values >0.05). Using logistic regression, adjusted and unadjusted 8-isoprostanes levels were associated with 1-hydroxypyrene (p = 0.02 and p = 0.001 respectively) and estimated 24-hour urine sodium method (p = 0.003 and p<0.001 respectively). However, only unadjusted 8-OHdG was associated with 1-hydroxypyrene (p = 0.03) and age (p = 0.007). Estimated 24-hour urinary sodium is high among Zambians and it is associated with lipid but not DNA oxidative stress. High exposure to polycyclic aromatic hydrocarbons is also associated with oxidative stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)