Merck
The Hippo kinases LATS1 and 2 control human breast cell fate via crosstalk with ERα.
Nature (2017-01-10)
Adrian Britschgi, Stephan Duss, Sungeun Kim, Joana Pinto Couto, Heike Brinkhaus, Shany Koren, Duvini De Silva, Kirsten D Mertz, Daniela Kaup, Zsuzsanna Varga, Hans Voshol, Alexandra Vissieres, Cedric Leroy, Tim Roloff, Michael B Stadler, Christina H Scheel, Loren J Miraglia, Anthony P Orth, Ghislain M C Bonamy, Venkateshwar A Reddy, Mohamed Bentires-Alj
ZUSAMMENFASSUNG
Cell fate perturbations underlie many human diseases, including breast cancer. Unfortunately, the mechanisms by which breast cell fate are regulated are largely unknown. The mammary gland epithelium consists of differentiated luminal epithelial and basal myoepithelial cells, as well as undifferentiated stem cells and more restricted progenitors. Breast cancer originates from this epithelium, but the molecular mechanisms that underlie breast epithelial hierarchy remain ill-defined. Here, we use a high-content confocal image-based short hairpin RNA screen to identify tumour suppressors that regulate breast cell fate in primary human breast epithelial cells. We show that ablation of the large tumour suppressor kinases (LATS) 1 and 2 (refs 5, 6), which are part of the Hippo pathway, promotes the luminal phenotype and increases the number of bipotent and luminal progenitors, the proposed cells-of-origin of most human breast cancers. Mechanistically, we have identified a direct interaction between Hippo and oestrogen receptor-α (ERα) signalling. In the presence of LATS, ERα was targeted for ubiquitination and Ddb1-cullin4-associated-factor 1 (DCAF1)-dependent proteasomal degradation. Absence of LATS stabilized ERα and the Hippo effectors YAP and TAZ (hereafter YAP/TAZ), which together control breast cell fate through intrinsic and paracrine mechanisms. Our findings reveal a non-canonical (that is, YAP/TAZ-independent) effect of LATS in the regulation of human breast cell fate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-BMI-1-Antikörper, Klon F6, clone F6, Upstate®, from mouse
Sigma-Aldrich
Anti-LATS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution