Merck
  • Synthesis and free radical scavenging activity of a novel metabolite from the fungus Colletotrichum gloeosporioides.
Synthesis and free radical scavenging activity of a novel metabolite from the fungus Colletotrichum gloeosporioides.
Bioorganic & medicinal chemistry letters (2006-09-05)
Marienca Femenía-Ríos, Carlos M García-Pajón, Rosario Hernández-Galán, Antonio J Macías-Sánchez, Isidro G Collado
ZUSAMMENFASSUNG
A novel metabolite (-)-1 was isolated as its peracetylated derivative, (-)-2-(3',4'-diacetoxyphenyl)-3,4-diacetoxytetrahydrofuran (2), from a strain of the phytopathogenic fungus Colletotrichum gloeosporioides CECT 20122. The synthesis of (-)-1 was carried out by ring-closing metathesis of diene 6 and stereoselective dihydroxylation of a dihydrofuran derivative 7 as key steps. The tetraol (-)-1 showed free radical scavenging activity comparable to that of BHT, caffeic acid or protocatechuic acid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
3,4-Dihydroxy-benzoesäure, ≥97.0% (T)
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, purum, ≥99.0% (GC)
Supelco
3,5-Di-tert-butyl-4-hydroxytoluol, analytical standard
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, tested according to Ph. Eur.
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)