Merck
  • Exposure to organophosphate ester flame retardants and plasticizers during pregnancy: Thyroid endocrine disruption and mediation role of oxidative stress.
Exposure to organophosphate ester flame retardants and plasticizers during pregnancy: Thyroid endocrine disruption and mediation role of oxidative stress.
Environment international (2020-10-29)
Yiming Yao, Mengqi Li, Liyang Pan, Yishuang Duan, Xiaoyu Duan, Yongcheng Li, Hongwen Sun
ZUSAMMENFASSUNG
Organophosphate esters (OPEs) are widely used as flame retardants and plasticizers in consumer and industrial products. Human exposure to OPEs raises concerns due to their endocrine disruptive potentials. Till now, the effects of OPEs on thyroid hormones (THs) and the mediating role of oxidative stress in pregnant women have not been studied. In this study, prenatal urinary concentrations of OPE metabolites (mOPEs), levels of free triiodothyronine (FT3), free thyroxine (FT4), thyroid-stimulating hormone (TSH), and oxidative stress levels of 8-hydroxy-2-deoxy guanosine (8-OHdG) and malondialdehyde (MDA) were measured in pregnant women (n = 360) from a coastal urbanized region and moderate socioeconomic status. Neonatal TSH in heel blood was also measured in newborns (n = 309). Dibutyl phosphate (DBP) and diphenyl phosphate (DPHP) were extensively detected with a median creatinine-adjusted level of 0.19 μg/g and 0.66 μg/g, respectively, and the median of ∑mOPEs was 1.82 μg/g. DBP and DPHP were included in the analysis. The concentrations of DBP and DPHP were positively associated with either maternal or neonatal TSH levels, while not for maternal FT3 and FT4 levels. Positive associations for maternal and neonatal TSH were particularly observed in girls as stratified by newborn sex suggesting a sex-selective difference. Furthermore, 8-OHdG, the biomarker of DNA damage, was found to be a major mediator (>60%) for the association between neonatal TSH and DPHP, suggesting that DNA damage is involved in fetal thyroid function disruption. On the other hand, MDA showed a partially suppressing effect (<40%) for the associations between mOPEs and neonatal TSH, which needs further clarification. For maternal TSH, both 8-OHdG and MDA showed moderate mediating effects while the direct effects of mOPEs on maternal TSH also contributed. These results suggest thyroid disrupting effects of OPE exposure on mothers and fetuses during pregnancy and the potential influence mediated by the oxidative stresses of DNA damage and lipid peroxidation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diphenylphosphat, 99%
Sigma-Aldrich
Dibutyl-phosphat, ≥97.0% (T)