Merck
  • Hexylitaconic acid: a new inhibitor of p53-HDM2 interaction isolated from a marine-derived fungus, Arthrinium sp.
Hexylitaconic acid: a new inhibitor of p53-HDM2 interaction isolated from a marine-derived fungus, Arthrinium sp.
Bioorganic & medicinal chemistry letters (2005-10-26)
Sachiko Tsukamoto, Takushi Yoshida, Hidetaka Hosono, Tomihisa Ohta, Hideyoshi Yokosawa
ZUSAMMENFASSUNG
A new inhibitor of p53-HDM2 interaction was isolated from a culture of marine-derived fungus, Arthrinium sp. The structure was identified to be (-)-hexylitaconic acid (1) by spectroscopic analysis. The inhibition of p53-HDM2 binding was tested by the ELISA method, and 1 inhibited the binding with an IC(50) value of 50 microg/mL. Although a number of synthetic inhibitors of p53-HDM2 interaction have been reported so far, 1 is the second inhibitor isolated from natural resources.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Itaconsäure, ≥99%
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)