- Synthesis of human long-term metabolites of dehydrochloromethyltestosterone and oxymesterone.
Synthesis of human long-term metabolites of dehydrochloromethyltestosterone and oxymesterone.
Steroids (2020-08-30)
Nicolas Kratena, Sandra Pfeffer, Valentin S Enev, Günter Gmeiner, Peter Gärtner
PMID32860782
ZUSAMMENFASSUNG
We herein report the synthesis of the long-term metabolites "M4" (IUPAC: 4-chloro-17-hydroxymethyl-17-methyl-18-norandrosta-4,13-dien-3-ol) of dehydrochloromethyl-testosterone (DHCMT, Oral Turinabol) and "Oxy M9" (4-hydroxy-17β-hydroxymethyl-17α-methyl-18-norandrosta-4,13-dien-3-one) of oxymesterone (Oranabol). Both compounds were derived from a common synthetic route starting from dehydroepiandrosterone acetate. Four different stereoisomers were evaluated for metabolite M4. The previously assigned structure could be corrected regarding the C-3 and C-17 stereocenters.
MATERIALIEN