Merck
  • A nitrogen-containing 3-alkyl-1,4-benzoquinone and a gomphilactone derivative from Embelia ribes.
A nitrogen-containing 3-alkyl-1,4-benzoquinone and a gomphilactone derivative from Embelia ribes.
Journal of natural products (2006-11-28)
Pengcheng Lin, Shuai Li, Sujuan Wang, Yongchun Yang, Jiangong Shi
ZUSAMMENFASSUNG
An unusual nitrogen-containing 3-alkyl-1,4-benzoquinone derivative, N-(3-carboxylpropyl)-5-amino-2-hydroxy-3-tridecyl-1,4-benzoquinone (1), and a gomphilactone derivative, 5,6-dihydroxy-7-tridecyl-3-[4-tridecyl-3-hydroxy-5-oxo-2(5H)-furylidene]-2-oxo-3(2H)-benzofuran (2), together with 14 known compounds, as well as the common plant metabolites sitosterol and daucosterol, were isolated from the ethanolic extract of the roots of Embelia ribes. Their structures were elucidated by means of spectroscopic methods. Cytotoxicities of the purified compounds were tested.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Supelco
5-Pentadecyl-resorcin, analytical standard