Merck
  • Downregulation of microRNA-605 indicates poor prognosis and promotes the progression of osteosarcoma.
Downregulation of microRNA-605 indicates poor prognosis and promotes the progression of osteosarcoma.
Oncology letters (2020-11-07)
Xiuling Yi, Chunlei Liu
ZUSAMMENFASSUNG
Osteosarcoma (OS) is a type of primary bone tumor, which is one of the leading causes of cancer-related death. MicroRNA (miR)-605 has been demonstrated to act as a prognostic biomarker and therapeutic target in various cancers, such as breast cancer and non-small cell lung cancer, but its function in OS remains unclear. The aim of the present study was to investigate the prognostic value of miR-605 in patients with OS by evaluating its expression levels and to explore the biological function of miR-605 in OS progression. For this purpose, tumor tissues and adjacent normal tissues were collected from OS patients, and the expression of miR-605 in the collected tissues and OS MG63, U2OS, HOS, and SAOS-2 cell lines was detected by quantitative real-time PCR. The prognostic value of miR-605 was evaluated by Kaplan-Meier survival curves and Cox regression analysis. The effects of miR-605 on OS cell proliferation, migration and invasion were analyzed by the CCK-8 and transwell assays, respectively. The results of the present study revealed that miR-605 was significantly downregulated in OS tissues compared with adjacent normal tissues, which was associated with the clinical stage and distant metastasis of patients. Additionally, the downregulation of miR-605 predicted the poor prognosis of patients with OS and served as an independent prognostic indicator. The downregulation of miR-605 enhanced cell proliferation, migration, and invasion of OS cells, which suggested that miR-605 may be involved in the progression of OS. The findings of the present study provide a new therapeutic target for the treatment of patients with OS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder