Merck
Flavan-4-ol Glycosides from the Rhizomes of Abacopteris penangiana.
Journal of natural products (2006-02-28)
Zhongxiang Zhao, Jinlan Ruan, Jing Jin, Jian Zou, Daonian Zhou, Wei Fang, Fanbo Zeng
ZUSAMMENFASSUNG
Four new flavan-4-ol glycosides, abacopterins A-D (1-4), were isolated from a methanol extract of the rhizomes of Abacopteris penangiana, together with two known compounds, triphyllin A (5) and 6,8-dimethyl-7-hydroxy-4'-methoxyanthocyanidin-5-O-beta-D-glucopyranoside (6). Their structures were elucidated by extensive spectroscopic analysis and chemical methods. The cytotoxic activity of 1-5 against HepG2 human hepatoma cells was investigated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications