Merck
  • Impact of chorionic somatomammotropin RNA interference on uterine blood flow and placental glucose uptake in the absence of intrauterine growth restriction.
Impact of chorionic somatomammotropin RNA interference on uterine blood flow and placental glucose uptake in the absence of intrauterine growth restriction.
American journal of physiology. Regulatory, integrative and comparative physiology (2020-11-05)
Amelia R Tanner, Cameron S Lynch, Asghar Ali, Quinton A Winger, Paul J Rozance, Russell V Anthony, Amelia R Tanner, Cameron S Lynch, Asghar Ali, Quinton A Winger, Paul J Rozance, Russell V Anthony
ZUSAMMENFASSUNG
Chorionic somatomammotropin (CSH) is one of the most abundantly produced placental hormones, yet its exact function remains elusive. Near-term [135 days of gestational age (dGA)], CSH RNA interference (RNAi) results in two distinct phenotypes: 1) pregnancies with intrauterine growth restriction (IUGR), and 2) pregnancies with normal fetal and placental weights. Here, we report the physiological changes in CSH RNAi pregnancies without IUGR. The trophectoderm of hatched blastocysts (9 dGA) were infected with lentiviral-constructs expressing either a scrambled control (Control RNAi) or CSH-specific shRNA (CSH RNAi), prior to transfer into synchronized recipient ewes. At 126 dGA, Control RNAi (n = 6) and CSH RNAi (n = 6) pregnancies were fitted with maternal and fetal catheters. Uterine and umbilical blood flows were measured at 132 dGA and nutrient uptakes were calculated by the Fick's principle. Control RNAi and CSH RNAi pregnancies were compared by analysis of variance, and significance was set at P ≤ 0.05. Absolute (mL/min) and relative (mL/min/kg fetus) uterine blood flows were reduced (P ≤ 0.05) in CSH RNAi pregnancies, but umbilical flows were not impacted. The uterine artery-to-vein glucose gradient (mmol/L) was significantly (P ≤ 0.05) increased. The uteroplacental glucose uptake (μmoL/min/kg placenta) was increased (P ≤ 0.05), whereas umbilical glucose uptake (μmoL/min/kg fetus) was reduced. Our results demonstrate that CSH RNAi has significant physiological ramifications, even in the absence of IUGR, and comparing CSH RNAi pregnancies exhibiting both IUGR and non-IUGR phenotypes may help determine the direct effects of CSH and its potential impact on fetal development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-GLUT-1-Antikörper, CT, from rabbit, purified by affinity chromatography