Merck
Vinculin and cell-cell adhesion.
Cell adhesion and communication (1998-06-25)
A Tozeren, S Wu, B Hoxter, W Xu, E D Adamson, S W Byers
ZUSAMMENFASSUNG
Vinculin, a 117-kDa protein, is a constituent of adhesion plaques and adherence junctions in non-muscle cells. We investigated the role of vinculin on the physical strength of cell-cell adhesion by conducting disaggregation assays on aggregates of parental wild-type F9 mouse embryonal carcinoma cells (clone BIM), two vinculin-depleted F9 cell lines, gamma 227 and gamma 229, and a reconstituted gamma 229 cell line (R3) that re-express vinculin. Immunoblotting demonstrated that the four cell lines used in the study had similar expressions of the cell-cell adhesion molecule E-cadherin and associated membrane proteins alpha- and beta-catenin. Double immunofluorescence analysis showed that, in contrast to the vinculin-null cell lines. BIM and R3 cells expressed abundant vinculin at the cell margins in adhesion plaques and in cell-cell margins that also contained actin. Laminar flow assays showed that both the vinculin-positive and vinculin-negative cell aggregates that were formed in culture in the course of 24 to 48 hours largely remained intact despite the imposition of shear flow at high shear rates. Since laminar flow imposed on cell aggregates act to separate cells from each other, our data indicate that F9 cells that were adherent to a substrate formed strong cell-cell adhesion bonds independent of vinculin expression. On the other hand, aggregates of vinculin-depleted gamma 229 and gamma 227 cells that were formed in suspension during a two-hour static incubation at 37 degrees C were desegregated more easily with the imposition of shear flow than the BIM and R3 cell aggregates formed under identical conditions. Loss of vinculin was associated with a reduction in cell-cell adhesion strength only among those cells lacking contact to a substrate. Overall, the results indicate that vinculin is not needed for forming strong cell-cell adhesion bonds between neighboring carcinoma cells which are adherent to the basal lamina.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Catenin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-α-Catenin antibody produced in rabbit, whole antiserum