Merck
  • Characterization of immune response induced against catalytic domain of botulinum neurotoxin type E.
Characterization of immune response induced against catalytic domain of botulinum neurotoxin type E.
Scientific reports (2020-08-20)
Priyanka Sonkar, Vinita Chauhan, Ritika Chauhan, Nandita Saxena, Ram Kumar Dhaked
ZUSAMMENFASSUNG
Botulinum neurotoxins (BoNTs) represent a family of bacterial toxins responsible for neuroparalytic disease 'botulism' in human and animals. Their potential use as biological weapon led to their classification in category 'A' biowarfare agent by Centers for Disease Control and Prevention (CDC), USA. In present study, gene encoding full length catalytic domain of BoNT/E-LC was cloned, expressed and protein was purified using Ni-NTA chromatography. Humoral immune response was confirmed by Ig isotyping and cell-mediated immunity by cytokine profiling and intracellular staining for enumeration of IFN-γ secreting CD4+ and CD8+ T cells. Increased antibody titer with the predominance of IgG subtype was observed. An interaction between antibodies produced against rBoNT/E-LC was established that showed the specificity against BoNT/E in SPR assay. Animal protection with rBoNT/E-LC was conferred through both humoral and cellular immune responses. These findings were supported by cytokine profiling and flow cytometric analysis. Splenocytes stimulated with rBoNT/E-LC showed a 3.27 and 2.8 times increase in the IFN-γ secreting CD4+ and CD8+ T cells, respectively; in immunized group (P < 0.05). Protection against BoNT/E challenge tended to relate with increase in the percentage of rBoNT/E-LC specific IL-2 in the splenocytes supernatant (P = 0.034) and with IFN-γ-producing CD4+ T cell responses (P = 0.045). We have immunologically evaluated catalytically active rBoNT/E-LC. Our results provide valuable investigational report for immunoprophylactic role of catalytic domain of BoNT/E.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-polyhistidin-Antikörper in Maus hergestellte Antikörper, clone HIS-1, ascites fluid
Sigma-Aldrich
Mouse Monoclonal Antibody Isotyping Reagents, sufficient for 1000 tests (clones) (by ELISA), sufficient for 40 tests (clones) (by immunodiffusion, ODD)