Merck
  • Synthesis and biological evaluation of N-pyrazolyl-N'-alkyl/benzyl/phenylureas: a new class of potent inhibitors of interleukin 8-induced neutrophil chemotaxis.
Synthesis and biological evaluation of N-pyrazolyl-N'-alkyl/benzyl/phenylureas: a new class of potent inhibitors of interleukin 8-induced neutrophil chemotaxis.
Journal of medicinal chemistry (2007-07-05)
Olga Bruno, Chiara Brullo, Francesco Bondavalli, Silvia Schenone, Angelo Ranise, Nicoletta Arduino, Maria B Bertolotto, Fabrizio Montecucco, Luciano Ottonello, Franco Dallegri, Massimiliano Tognolini, Vigilio Ballabeni, Simona Bertoni, Elisabetta Barocelli
ZUSAMMENFASSUNG
Neutrophils chemotaxis is a complex multistep process that, if upregulated, causes acute inflammation and a number of autoimmune diseases. We report here the synthesis of a new N-(4-substituted)pyrazolyl-N'-alkyl/benzyl/phenylureas that are potent inhibitors of interleukin-8 (IL8)-induced neutrophil chemotaxis. The first series of compounds, obtained by functionalization with a urea moiety of the 5-amino-1-(2-hydroxy-2-phenylethyl)-1H-pyrazole-4-carboxylic acid ethyl ester 3, blocked the IL8-induced neutrophil chemotaxis, while they did not block N-formylmethionylleucylphenylalanine-mediated chemotaxis. The most active compounds, 3-benzyl- (4d), 3-(4-benzylpiperazinyl)- (4i), 3-phenyl- (4k) and 3-isopropylureido (4a) derivatives, showed an IC50 of 10, 14, 45, and 55 nM, respectively. Several different molecules were then synthesized to obtain more information for SAR study. Compounds 4a, 4d, and 4k were inactive in the binding assays on CXCR1 and CXCR2 (IL8 receptors), whereas they inhibited the phosphorylation of PTKs (protein tyrosine kinases) in the 50-70 kDa region. Moreover, in the presence of the same derivatives, we observed a complete block of F-actin rise and pseudopod formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.