Merck
  • High-Performance Blue Perovskite Light-Emitting Diodes Enabled by Efficient Energy Transfer between Coupled Quasi-2D Perovskite Layers.
High-Performance Blue Perovskite Light-Emitting Diodes Enabled by Efficient Energy Transfer between Coupled Quasi-2D Perovskite Layers.
Advanced materials (Deerfield Beach, Fla.) (2020-11-21)
Zhenwei Ren, Jiahao Yu, Zhaotong Qin, Jing Wang, Jiayun Sun, Christopher C S Chan, Shihao Ding, Kai Wang, Rui Chen, Kam Sing Wong, Xinhui Lu, Wan-Jian Yin, Wallace C H Choy
ZUSAMMENFASSUNG
While there has been extensive investigation into modulating quasi-2D perovskite compositions in light-emitting diodes (LEDs) for promoting their electroluminescence, very few reports have studied approaches involving enhancement of the energy transfer between quasi-2D perovskite layers of the film, which plays very important role for achieving high-performance perovskite LEDs (PeLEDs). In this work, a bifunctional ligand of 4-(2-aminoethyl)benzoic acid (ABA) cation is strategically introduced into the perovskite to diminish the weak van der Waals gap between individual perovskite layers for promoting coupled quasi-2D perovskite layers. In particular, the strengthened interaction between coupled quasi-2D perovskite layers favors an efficient energy transfer in the perovskite films. The introduced ABA can also simultaneously passivate the perovskite defects by reducing metallic Pb for less nonradiative recombination loss. Benefiting from the advanced properties of ABA incorporated perovskites, highly efficient blue PeLEDs with external quantum efficiency of 10.11% and a very long operational stability of 81.3 min, among the best performing blue quasi-2D PeLEDs, are achieved. Consequently, this work contributes an effective approach for high-performance and stable blue PeLEDs toward practical applications.