Merck
  • Identifying merozoite surface protein 4 and merozoite surface protein 7 Plasmodium falciparum protein family members specifically binding to human erythrocytes suggests a new malarial parasite-redundant survival mechanism.
Identifying merozoite surface protein 4 and merozoite surface protein 7 Plasmodium falciparum protein family members specifically binding to human erythrocytes suggests a new malarial parasite-redundant survival mechanism.
Journal of medicinal chemistry (2007-10-20)
Yesid Garcia, Alvaro Puentes, Hernando Curtidor, Gladys Cifuentes, Claudia Reyes, Jose Barreto, Armando Moreno, Manuel E Patarroyo
ZUSAMMENFASSUNG
Plasmodium falciparum merozoite surface proteins (MSP-1 to -11) have been involved in merozoite interaction with the red blood cell (RBC) surface. Peptides covering complete MSP-4 and MSP-7 amino acid sequences were synthesized and tested in RBC binding assays. One MSP-4 high activity binding peptide (HABP) and five MSP-7 HABPs were found having specific binding to RBC surface. MSP-4 and MSP-7 HABP binding was sensitive to enzymatic treatment; they recognized a 52 kDa erythrocyte membrane protein. MSP-4 HABP had low invasion inhibition, suggesting it might bind to RBCs and also be involved in physiological mechanisms, while MSP-7 HABPs displayed different invasion inhibition activity (83-24%) in in vitro tests, suggesting different roles for both proteins during invasion. Structural characteristics found when comparing the MSP-4 HABP with MSP-HABPs displaying epidermal growth factor-like sequences suggested that these redundant MSP-family proteins could be a new parasite strategy for evading host genetic variability and immune pressure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioUltra, ≥99.0% (T)